missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TIRAP (TLR2 and TLR4) Inhibitor Peptide Set
Shop All Bio Techne Products
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
TLR2 / TLR4 Inhibitor Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKKPGFLRDPWCKYQML (Inhibitor sequence: PGFLRDPWCKYQML); Molecular weight: 4097.92 Da ; Antennapedia Control Peptide: 1mg (lyophilized); sequence: DRQIKIWFQNRRMKWKK; Molecular weight: 2361 Da
Specifications
Specifications
| Color | White |
| Content And Storage | Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | Inhibit TLR2 and TLR4 signaling |
| Product Type | TIRAP (TLR2 and TLR4) Inhibitor Peptide Set |
| Quantity | 5 mg |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction