Learn More
Elma™ Elma Dry
Click to view available options
Model:
TD 120
TD 30
TD 300
Unit Size:
Each
Description
Elma Dry units are available in three sizes (30, 120 and 300). They are designed for use in production facilities, workshops and service areas.
- After cleaning in the ultrasonic bath, the items in the Elma™ basket are first rinsed in the Elma™ Sink and then dried easily and conveniently in the Elma Dry
- Cold/hot-air blower (70°C) ensures that even delicate parts can be dried gently
- In cold-air mode, the parts can also be cooled down quickly
- Cover includes ventilation slits that maximize the air turbulence and discharge the humid air to the outside while reducing energy consumption
- Dust filter on the underside of the base is replaceable and prevents recontamination of the cleaned parts due to dust in the room air
- Due to the safety shutdown when the limit temperature is reached, Elma Dry units are both safe and gentle on their contents
Specifications
Specifications
| Type | Elma Dry |
| For Use With (Equipment) | Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |
| Model | TD 300 |