Learn More
BD ELISPOT Sets
Product Code. 15826518
Change view
Click to view available options
Antigen:
Granzyme B
IFN-gamma
IL-10
IL-2
IL-4
IL-5
TNF
Cross Reactivity:
Human
Mouse
Product Type:
Human Granzyme B ELISPOT set
Human IFN-gamma ELISPOT set
Human IL-10 ELISPOT set
Human IL-2 ELISPOT set
Human IL-4 ELISPOT set
Human IL-5 ELISPOT set
Human TNF ELISPOT set
Mouse IFN-gamma ELISPOT set
Mouse IL-2 ELISPOT set
Mouse IL-4 ELISPOT set
Mouse IL-5 ELISPOT set
Mouse TNF ELISPOT set
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Antigen Cross Reactivity Product Type Unit Size
15826518 IFN-gamma Mouse Mouse IFN-gamma ELISPOT set Each
15866708 IFN-gamma Human Human IFN-gamma ELISPOT set Each
15856508 IL-5 Mouse Mouse IL-5 ELISPOT set Each
15846518 IL-5 Human Human IL-5 ELISPOT set Each
15876668 TNF Mouse Mouse TNF ELISPOT set Each
15836518 IL-4 Human Human IL-4 ELISPOT set Each
15886828 Granzyme B Human Human Granzyme B ELISPOT set Each
15866508 IL-2 Mouse Mouse IL-2 ELISPOT set Each
15816658 TNF Human Human TNF ELISPOT set Each
15836478 IL-4 Mouse Mouse IL-4 ELISPOT set Each
15846478 IL-10 Human Human IL-10 ELISPOT set Each
15826598 IL-2 Human Human IL-2 ELISPOT set Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15826518 Supplier BD Supplier No. 551083
 Restricted Item
Estimated Shipment: 24-09-2026
Includes
Unlabeled capture antibody (no azide/low endotoxin format), Biotinylated detection antibody, Streptavidin-HRP, 10 ELISPOT plates, Certificate of Analysis providing lot-specific optimal reagent concentrations
Sufficient capture antibody, detection antibody, and streptavidin-horseradish peroxidase for 10 x 96 wells
  • High sensitivity
  • High throughput, high content analysis
  • Minimal volume of biological material required
  • Suitable for frozen/thawed biological samples
  • Compatibility with other assays—cells analyzed by BD ELISPOT Sets can be used for cloning, proliferation assays, flow cytometry, or other methods of analysis
TRUSTED_SUSTAINABILITY
Specifications
Antigen IFN-gamma
Cross Reactivity Mouse
Includes Unlabeled capture Antibody, detection Antibody, Streptavidin HRP, 10 ELISPOT plates
Quantity 10 x 96 Tests
Content And Storage 4°C
For Use With (Application) Cloning, proliferation assays, flow cytometry or other methods of analysis
Product Type Mouse IFN-gamma ELISPOT set