Learn More
Skytec Elements™ Elements EMG201 Safety Gloves
Product Code. p-200110061
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
10 pairs
Product Code. 30274637
Brand: Skytec Elements™ EMG00201FD
Only null left
Add to basket
Add to basket
Fully coated nitrile with a nitrile sandy finish palm. Excellent grip in wet and oily conditions as well as oil and water submersion.
  • Multi-Purpose Polyester Fully Coated Nitrile Double Dipped Glove
  • Designed for easy movement and continuous wear
  • Elasticated wrist provides a secure and comfortable fit
  • Seamless liner prevents irritation
Specifications
ANSI Abrasion Rating ANSI 4
Construction Seamless Knit
Coating Material Nitrile
Color Gray, Black
Exterior Finish Sandpatch
Hand Ambidextrous
Latex Free Yes
Material Polyester
Product Type General Purpose Gloves
Size Small
Liner Material Polyester
Length (Metric) 250 mm