Learn More
Bibby Scientific™ Electrothermal™ Heating Element for EM/EME
Product Code. 11876475
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
5000 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11876475 250 mL Each
11738412 1000 mL Each
11748412 2000 mL Each
11768412 5000 mL Each
11728412 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11876475 Supplier Bibby Scientific™ Supplier No. 0301235
  • ONLINE EXCLUSIVE PRICE
    720.00€
    499.02€ / Pack of 1
    Save 220.98€
    30% Off
Estimated Shipment: 29-12-2026
Bibby Scientific Electrothermal Heating Element for EM/EME, Capacity: 250 mL, For Use With: 14314 EM Series Heating Mantles, Voltage: 230 V, Frequency: 50/60 Hz
Specifications
Capacity (Metric) 250 mL
For Use With 14314 EM Series Heating Mantles
Voltage 230 V
Product Type Heating Element
Frequency 50/60 Hz