Learn More
Honeywell™ ELECTROSOFT Class 1 Latex Gloves
Product Code. 12677573
Click to view available options
Size:
8
9
10
Unit Size:
Each
Product Code. 12677573
Brand: Honeywell™ 209191208
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Honeywell™ ELECTROSOFT Class 1 Latex Gloves are ideal for electrical work and should be used with leather protectors for mechanical protection.
  • Beige natural latex insulating glove
  • Cut edge
  • Category RC
  • Ergonomic shape and lightly powdered interior make the glove easy to slip on and off
  • The natural latex base offers high dielectric characteristics
  • Its two-toned composition, with its beige interior/black exterior, indicates wear on the whole of the glove
  • Its thickness provides good dexterity while offering protection up to 7500 volts
  • To be used with leather protectors for mechanical protection
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 60903-2003, CEI 903-2002
Coating Material Latex
Color Beige
Size 8
Thickness (Metric) 1.5 mm
Length (Metric) 36 cm