Learn More
Thermo Scientific™ Electroporation Cuvettes
GREENER_CHOICE
Product Code. p-4524565 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Width (Metric) Gap:
1 mm
2 mm
4 mm
Unit Size:
Pack of 50
Product Code. Width (Metric) Gap unitSize
12338212 1 mm Pack of 50
10227570 2 mm Pack of 50
12348212 4 mm Pack of 50
3 options
Product Code. 12338212
Brand: Thermo Scientific™ 551011
Edge
Increase the ease of sample identification and labelling with these premium-grade polycarbonate electroporation cuvettes, which have frosted, colored caps.
Increase the ease of sample identification and labeling with premium-grade polycarbonate Thermo Scientific™ Electroporation Cuvettes, which have frosted, colored caps.
  • Available in 1mm, 2mm, or 4mm gap sizes
  • Pre-sterilized and individually wrapped
  • Compatible with all common electroporators
TRUSTED_SUSTAINABILITY
Specifications
Type Retort Ring
Material Stainless Steel
Width (Metric) Gap 1 mm
Diameter (Metric) 50 mm