Learn More
3M™ Electronic Test System Data Reader
Product Code. 17781669 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
ETS Sensing Unit (4308)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17781669 ETS Sensing Unit (4308) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17781669 Supplier 3M™ Supplier No. 7100194284
  • ONLINE EXCLUSIVE PRICE
    556.89€
    434.34€ / Pack of 1
    Save 122.55€
    22% Off
Estimated Shipment: 12-01-2027
The ETS Data Reader reads data from your main ETS unit and downloads to a PC.
  • Service parts available to authorized entities only
  • Uses LED wireless connection to read data from ETS
  • Accessory to main ETS Sensing Unit
Specifications
For Use With (Application) Reading Data From ETS Unit
For Use With (Equipment) ETS Sensing Unit (4308)
Model 4309