Learn More
Macherey-Nagel™ Electronic Decapper
Click to view available options
For Use With (Equipment):
For 11mm Aluminum Crimp Caps
For 20mm Aluminum Crimp Caps
Unit Size:
Each
Specifications
Specifications
| No. per Pack | 1 Each |
| Type | Electronic Decapper |
| For Use With (Equipment) | For 11mm Aluminum Crimp Caps |