Learn More
Mettler Toledo™ Electrolyte For Thiocyanate ISE
Product Code. 11997711 Shop All Mettler Toledo Products
Change view
Click to view available options
Quantity:
20 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11997711 20 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11997711 Supplier Mettler Toledo™ Supplier No. 51107872
Specifications
Description Electrolyte for Thiocyanate ISE, 20mL
Filling Solution Type Electrolyte
Packaging 20 mL
Quantity 20 mL