Learn More
BINDER™ Electric Access Port
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
VDL 115, VDL 53, VDL 23
Unit Size:
Each
Description
- 9-pin
- Vacuum-tight access port
- For transmitting electrical currents, voltages, or signals (max. 50 V / 2 A per pin; vacuum connection DN 16 ISO-KF)
Specifications
Specifications
| Description | Electric access port - 9-pin vacuum-tight access port for measuring lines |
| For Use With (Equipment) | VDL 115, VDL 53, VDL 23 |
| Material | Silicone |
| Type | Access Port |
| Electrical Requirements | 50 V, 2 A / pin |