Learn More
BINDER™ Electric Access Port
Product Code. 16272011 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
VDL 115, VDL 53, VDL 23
Unit Size:
Each
Product Code. 16272011
Brand: BINDER™ 80121238
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For Use With: VDL 115, VDL 53, VDL 23
  • 9-pin
  • Vacuum-tight access port
  • For transmitting electrical currents, voltages, or signals (max. 50 V / 2 A per pin; vacuum connection DN 16 ISO-KF)
Specifications
Description Electric access port - 9-pin vacuum-tight access port for measuring lines
For Use With (Equipment) VDL 115, VDL 53, VDL 23
Material Silicone
Type Access Port
Electrical Requirements 50 V, 2 A / pin