missing translation for 'onlineSavingsMsg'
Learn More

PL BioScience ELAREM™ Perform-FD PLUS, Fibrinogen-depleted Human Platelet Lysate, No Heparin Added or Required

Product Code. p-200008740
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
This item is not returnable. View return policy

Product Code. 17977741

Brand: PL BioScience PE30611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELAREM™ Perform-FD PLUS Research Grade is a fibrinogen-depleted and heparin-free Human Platelet Lysate of EU origin. As a xeno-free, clot-free and easy-to-use cell culture supplement, Perform-FD PLUS supports the efficient and safe in vitro expansion of various primary cells.

  • Clot-free and Fibrinogen-depleted HPL: ELAREM™ Perform-FD PLUS is a fibrinogen-depleted, ready-to-use Human Platelet Lysate that is widely applicable.
  • No Heparin Added or Required: Due to the unique fibrinogen-depletion technique, ELAREM™ Perform-FD PLUS is clot-free and heparin-free. The product neither requires the addition of anticoagulants (e.g. heparin) in cell culture nor during manufacturing.
  • High Performance Lab Process: ELAREM™ Perform-FD PLUS efficiently and safely supports in vitro expansion of various primary cells and cell lines.
  • EU Origin for Long Traceability: ELAREM™ Perform-FD PLUS is manufactured from platelet units obtained from healthy blood donors at licensed blood centers in the EU. ELAREM™ Perform-FD PLUS is derived from EU origin platelet units which ensures long raw material traceability periods.
  • Intended Use: ELAREM™ Perform-FD PLUS is for in vitro and manufacturing use only. The product is not intended for direct therapeutic use. For safety precautions, please see appropriate Safety Data Sheet (SDS).

Specifications

Color Yellow
Culture Type Adherent Cell Culture, Suspension Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Sterility Sterile
Sterilization Method Sterile-filtered
Formulation Liquid, Human Platelet Lysate, Fibrinogen-depleted
Purity or Quality Grade Research Grade
Quantity 50 mL
Shipping Condition Dry Ice
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.