Learn More
Elaiophylin (>85%), TRC
Specifications
Specifications
| Chemical Name or Material | Elaiophylin |
| CAS | 37318-06-2 |
| Molecular Formula | C54H88O18 |
| Quantity | 2.5 mg |
| Synonym | 5001B, Antibiotic 5001B, Antibiotic 56-62, Antibiotic A 164AB, Azalomycin B, Efomycin E, Efomycine E, Gopalamicin, [7S-[3E,5E,7R*,8R*[1R*,2S*,3R*(2S*,4S*,5S*,6S*)],11E,13E,15R*,16R*[1R*,2S*,3R*(2S*,4S*,5S*,6S*)]]]-8,16-Bis[3-[4-[(2,6-dideoxy-alpha-L-lyxo-hexopyranosyl)oxy]-5-ethyltetrahydro-2-hydroxy-6-methyl-2H-pyran-2-yl]-2-hydroxy-1-methylbutyl]-7,15-dimethyl-1,9-dioxacyclohexadeca-3,5,11,13-tetraene-2,10-dione, [7S-[3E,5E,7R*,8R*[1R*,2S*,3R*(2S*,4S*,5S*,6S*)],11E,13E,15R*,16R*[1R*,2S*,3R*(2S*,4S*,5S*,6S*)]]]-8,16-Bis[3-[4-[(2,6-dideoxy-alpha-L-lyxo-hexopyranosyl)oxy]-5-ethyltetrahydro-2-hydroxy-6-methyl-2H-pyran-2-yl]-2-hydroxy-1-methylbutyl]-7,15-dimethyl-1,9-dioxacyclohexadeca-3,5,11,13-tetraene-2,10-dione, 1,9-Dioxacyclohexadecane, elaiophylin deriv., (+)-Elaiophylin |
| InChI Formula | InChI=1S/C54H88O18/c1-13-37-33(9)71-53(63,25-41(37)67-45-23-39(55)49(61)35(11)65-45)31(7)47(59)29(5)51-27(3)19-15-17-22-44(58)70-52(28(4)20-16-18-21-43(57)69-51)30(6)48(60)32(8)54(64)26-42(38(14-2)34(10)72-54)68-46-24-40(56)50(62)36(12)66-46/h15-22,27-42,45-52,55-56,59-64H,13-14,23-26H2,1-12H3/b19-15+,20-16+,21-18+,22-17+/t27-,28-,29-,30-,31-,32-,33+,34+,35-,36-,37+,38+,39-,40-,41+,42+,45-,46-,47+,48+,49+,50+,51-,52-,53+,54+/m0/s1 |
| SMILES | CC[C@@H]1[C@@H](C)O[C@](O)(C[C@H]1O[C@H]2C[C@H](O)[C@H](O)[C@H](C)O2)[C@@H](C)[C@H](O)[C@H](C)[C@H]3OC(=O)\C=C\C=C\[C@H](C)[C@H](OC(=O)\C=C\C=C\[C@@H]3C)[C@@H](C)[C@@H](O)[C@H](C)[C@@]4(O)C[C@@H](O[C@H]5C[C@H](O)[C@H](O)[C@H](C)O5)[C@H](CC)[C@@H](C)O4 |
| IUPAC Name | (3E,5E,7S,8S,11E,13E,15S,16S)-8,16-bis[(2S,3R,4S)-4-[(2R,4R,5R,6R)-4-[(2R,4S,5S,6S)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-5-ethyl-2-hydroxy-6-methyloxan-2-yl]-3-hydroxypentan-2-yl]-7,15-dimethyl-1,9-dioxacyclohexadeca-3,5,11,13-tetraene-2,10-dione |
| Molecular Weight (g/mol) | 1025.27 |
| Formula Weight | 1024.5971 |