Learn More
UVP EL Lamps
Product Code. 11778251
Change view
Click to view available options
Wavelength:
254 nm
302 nm
365 nm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength unitSize
11778251 254 nm Each
11738251 302 nm Each
11758241 365 nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11778251 Supplier UVP Supplier No. 95020002
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Light Source UV, White Light
Voltage 230 V
Wattage 8 W
Wavelength 254 nm
Amperage 0.16 A
Dimensions (L x W x H) 376 x 97 x 64 mm
Electrical Requirements 8 W, 230 V, 50/60 Hz
Height (Metric) 64 mm
Hertz 50/60 Hz
Length (Metric) 376 mm
Model EL Series Lamp
Packaging Quantity 1 Ea.
Width (Metric) 97 mm
Product Type UV Lamp