missing translation for 'onlineSavingsMsg'
Learn More

Eicosanedial, TRC

Product Code. 30380601
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380601 10 mg 10mg
30382553 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30380601 Supplier Toronto Research Chemicals Supplier No. TRCE33203010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Eicosanedial
CAS 1951435-37-2
Molecular Formula C20 H38 O2
Quantity 10 mg
InChI Formula InChI=1S/C20H38O2/c21-19-17-15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-16-18-20-22/h19-20H,1-18H2
SMILES O=CCCCCCCCCCCCCCCCCCCC=O
IUPAC Name icosanedial
Molecular Weight (g/mol) 310.514
Formula Weight 310.287
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.