missing translation for 'onlineSavingsMsg'
Learn More

Eflornithine Hydrochloride Monohydrate, TRC

Product Code. 30378247
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30378247

Brand: Toronto Research Chemicals TRCE436000250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Difluoromethylornithine hydrochloride monohydrate
CAS 96020-91-6
Molecular Formula C6 H12 F2 N2 O2 . Cl H . H2 O
Quantity 250 mg
Synonym Ornithine, 2-(difluoromethyl)-, hydrochloride, hydrate (1:1:1), DL-Ornithine, 2-(difluoromethyl)-, monohydrochloride, monohydrate, Ornithine, 2-(difluoromethyl)-, monohydrochloride, monohydrate (9CI), Eflornithine hydrochloride monohydrate
InChI Formula InChI=1S/C6H12F2N2O2.ClH.H2O/c7-4(8)6(10,5(11)12)2-1-3-9;;/h4H,1-3,9-10H2,(H,11,12);1H;1H2
SMILES O.Cl.NCCCC(N)(C(F)F)C(=O)O
IUPAC Name 2,5-diamino-2-(difluoromethyl)pentanoic acid;hydrate;hydrochloride
Molecular Weight (g/mol) 236.64
Formula Weight 236.0739
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.