missing translation for 'onlineSavingsMsg'
Learn More

Eflornithine Hydrochloride Monohydrate, TRC

Product Code. 30375811
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
This item is not returnable. View return policy

Product Code. 30375811

Brand: Toronto Research Chemicals TRCE4360002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Difluoromethylornithine hydrochloride monohydrate
CAS 96020-91-6
Molecular Formula C6 H12 F2 N2 O2 . Cl H . H2 O
Quantity 2.5 g
Synonym Ornithine, 2-(difluoromethyl)-, hydrochloride, hydrate (1:1:1), DL-Ornithine, 2-(difluoromethyl)-, monohydrochloride, monohydrate, Ornithine, 2-(difluoromethyl)-, monohydrochloride, monohydrate (9CI), Eflornithine hydrochloride monohydrate
InChI Formula InChI=1S/C6H12F2N2O2.ClH.H2O/c7-4(8)6(10,5(11)12)2-1-3-9;;/h4H,1-3,9-10H2,(H,11,12);1H;1H2
SMILES O.Cl.NCCCC(N)(C(F)F)C(=O)O
IUPAC Name 2,5-diamino-2-(difluoromethyl)pentanoic acid;hydrate;hydrochloride
Molecular Weight (g/mol) 236.64
Formula Weight 236.0739
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.