Learn More
Edwards Rotary Vane Vacuum Pump Fittings: Clamping Ring
Product Code. 10462141
Change view
Click to view available options
For Use With:
Rotary vane vacuum pump
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
10462141 Rotary vane vacuum pump Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10462141 Supplier Edwards Supplier No. C10514401
  • ONLINE EXCLUSIVE PRICE
    16.40€
    11.06€ / Pack of 1
    Save 5.34€
    32% Off
Estimated Shipment: 14-01-2027
Range of coupling clamps manufactured from high technology polymer for vacuum system builder
  • Clamping ring: compact stainless-steel
  • Polymer clamp: lightweight, neat appearance
  • Aluminum clamp: rugged Pneurop standard
  • Metal clamp: suitable for aluminum and indium seals — wide temperature range
  • Claw clamps and bolts: wide range optimized for many applications — high strength CF bolts for UHV flanges
Specifications
For Use With Rotary vane vacuum pump
Material Stainless Steel
Type Clamping Ring