Learn More
EDTA (Di Sodium Salt) Solution, EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA
Product Code. 12960924 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1000 mL
Concentration or Composition (by Analyte or Components):
0.01 mol/L
0.1 mol/L
Unit Size:
1000mL
Product Code. 12960924
Brand: Fisher Chemical VL 219 1000
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Quantity 1000 mL
Concentration or Composition (by Analyte or Components) 0.01 mol/L
Physical Form Liquid