missing translation for 'onlineSavingsMsg'
Learn More

Sartorius EDTA 0.5 M

Product Code. 31908491 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31908491 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908491 Supplier Sartorius Supplier No. 018621B
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

It is an enzyme-free, chemically defined, animal component-free concentrated dissociation solution.

  • It is a concentrated stock solution for the preparation of 0.5 mM EDTA dissociation solution.
  • It is suitable for the dissociation of human pluripotent stem cells (hESC and hiPSC).

Specifications

Classification Chemically-defined, Animal Origin-free
Form Liquid
Product Type EDTA
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.