Learn More
Brand™ Edisonite Super Universal Detergent
Product Code. 15528081 Shop All Brand Products
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
Product Code. 15528081
Brand: Brand™ 44361
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Product Line Edisonite™ SUPER
Quantity 1 kg
Volume (Metric) 1 kg
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : sodium carbonate, benzenesulphonic acid,C10-13 derivitives,sodium salt, fumaric acid