Learn More
BRAND™ Edisonite Super Universal Detergent
Product Code. 10363461 Shop All Brand Products
Click to view available options
Quantity:
1 Bucket
Unit Size:
5kg
Product Code. 10363461
Brand: BRAND™ 44365
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Neutral detergent. Powder compound for immersion and ultrasonic bath. Ideal for removal of natural fats and oils, proteins and blood. EDISONITE™ SUPER is recommended for optical glass, for plastics sensitive to alkalis and rubbers.
  • Powder compound for immersion and ultrasonic bath.
  • Ideal for removal of natural fats and oils, proteins and blood.
  • Recommended for optical glass, for plastics sensitive to alkalis and rubbers.
  • The actual Safety Data Sheet can be downloaded at www.brand.de
TRUSTED_SUSTAINABILITY
Specifications
Product Type Neutral Detergent
Form Powdered Compound
For Use With (Application) Removal of natural fats and oils, proteins and blood
For Use With (Equipment) Optical glass, for plastics sensitive to alkalis and rubbers
Quantity 1 Bucket
Volume (Metric) 5 kg