missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Edisonite™ Classic Universal Powdered Detergent

Product Code. 10120451 Shop All Brand Products
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 10120451

Brand: BRAND™ 44301

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Alkaline detergent

  • Light green powder compound for immersion and ultrasonic bath
  • Ideal for removal of natural fats and oils, proteins and blood
  • Durable protection against corrosion damage
  • Ingredients: > 30% phosphates, < 5% anionic surfactants, non-ionic surfactants, corrosion inhibitors, auxiliary agents
  • Concentration for use: 0.5 - 1% (5 - 10 g/L)
  • Bulk density: approx. kg/m³
  • pH value (0.5%): approx. 11.5
  • pH value (1%): approx. 11.5
  • Use on zinc and non-anodized aluminum is restricted
  • TRUSTED_SUSTAINABILITY

    Specifications

    Container Type Jar
    For Use With (Application) Ideal for removal of natural fats and oils, proteins and blood
    Quantity 1 kg
    Volume (Metric) 1 kg
    compliance-icons compliance-icons
    Product Identifier
    • alkaline detergent edisonite
    Signal Word
    • Danger
    Hazard Category
    • Serious eye damage/eye irritation Category 1
    • Skin corrosion/irritation Category 2
    • Specific target organ toxicity Category 3
    Hazard Statement
    • H315-Causes skin irritation.
    • H318-Causes serious eye damage.
    • H335-May cause respiratory irritation.
    Precautionary Statement
    • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
    • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P310-Immediately call a POISON CENTER/doctor/.
    Supplemental information
    • MIXTURE LIST-Contains : trisodiumorthophosphate, sodium dodecylsulphate
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.