Learn More
Ansell EDGE™ 48-125 Polyurethane Coated Polyester Gloves
Product Code. 16327721 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Product Code. 16327721
Brand: Ansell 48125070
Only null left
Add to basket
Edge
Add to basket
Polyurethane coating on the palm and fingers provides a high level of abrasion resistance, as well as enhanced grip capabilities.
  • EN level 4 of abrasion resistance
  • Tear resistance
  • Good grip for improved productivity
  • Shows dirt contamination quickly
  • Seamless structure to ensure comfort and encourage safety
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category ll; REACH Compliant
Construction Seamless Knit
Coating Material Polyurethane
Color White
Exterior Finish No Texture
Hand Specific
Material Polyester
Product Type Seamless Knit Gloves
Size 7
For Use With (Application) Inspection, General Handling, and Packaging
Glove Cuff Style Knit
Series 48-125