missing translation for 'onlineSavingsMsg'
Learn More

Ansell EDGE™ 14-662 Certified PVC-Coated Chemical Resistant Gloves - Safety Cuff

Product Code. 16609103 Shop All Ansell Products
Click to view available options
Size:
8
9
10
11
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 16609103

Brand: Ansell 14662110

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

High quality gloves provide reliable protection to workers who are exposed to oil, grease, caustics, and alcohols. Textured finish protects against abrasions while also enhancing grip capabilities in wet and oily conditions.

  • Protects workers that are exposed to oil, grease, acid, caustics and alcohols
  • Rough finish protects against abrasions while enhancing grip, even in wet and oily conditions
  • Flexibility and comfort leads to less hand fatigue and better worker efficiency
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 407:2004, EN 420:2003 + A1:2009, REACH Compliant
Applications Industrial
Color Blue
Exterior Finish Fully Textured
Latex Free Yes
Material PVC
Product Type Chemical-resistant Gloves
Size 11
EN388 Standard 3121A
EN ISO 374-1 Type C
EN ISO 374 - 5 Yes
Glove Cuff Style Safety
Series 14-662
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.