Learn More
Lauda LAUDA ECO RE 630 S Cooling thermostat 230 V, 50 HzEU plug
GREENER_CHOICE
Brand:  Lauda L001335
LAUDA ECO RE 630 S Cooling thermostat 230 V, 50 HzEU plug
Product Code. 15731377
  • ONLINE EXCLUSIVE PRICE
    6040.00€
    5720.00€ / Each
    Save 320.00€
    5% Off
Only null left
Estimated Shipment: 07-01-2026