Learn More
Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 25 mm Shelf Height
Eco AluRack for upright freezers is designed to hold microtiterplates.
Marca: Tenak™ TE15532
Dettagli aggiuntivi : Weight : 1.60000kg
Descrizione
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifica
25 mm | |
Upright Freezers | |
8 x 6 |
540 x 135 x 212 mm | |
Microtiterplates (48) | |
Aluminum |