Learn More
Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 25 mm Shelf Height
Eco AluRack for upright freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE15464
Additional Details : Weight : 1.60000kg
 View more versions of this product
Product Code. 12438230
  • 160.00€ / Each
Estimated Shipment: 04-10-2024
Description
  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of aluminum
Specifications
Specifications
25 mm
Upright Freezers
10 x 5
450 x 135 x 265 mm
Microtiterplates (50)
Aluminum