Learn More
Sharpsguard™ Eco 2.5 Waste Container
Product Code. 17764844
Change view
Click to view available options
Color:
Orange
Yellow
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17764844 Orange Each
17774844 Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17764844 Supplier Sharpsguard™ Supplier No. DD472OLR
  • ONLINE EXCLUSIVE PRICE
    29.23€
    1.59€ / Pack of 1
    Save 27.64€
    94% Off
Estimated Shipment: 15-12-2026
The new Sharpsguard™ eco containers, made using at least 40% recycled plastic that is used for the disposal of sharps that have not been contaminated with medicines.
  • Sharpsguard™ eco 2.5 is an ideal general-purpose sharps container, it provides maximum capacity in the minimum working space.
  • This container is supplied with the base and lid separately. Details about how to lock the container and keep the users and contents safe can be found on the label.
  • It is designed to fit safely and securely into our POUDS™ tray range.
  • This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-02-01, 18-02-02.
Specifications
Capacity (Metric) 2.21 L
Diameter (Metric) Outer 176 mm
Color Orange
Height (Metric) 166 mm
Quantity 1 Ea.
Weight (Metric) 232 g