Learn More
BTX™ ECM™ 830 Electrode Adapter Set Banana to Pin Tip
Click to view available options
Connection Type:
Banana to Pin Tip
Unit Size:
1 set
Description
- For connection to electroporation generators
Specifications
Specifications
| For Use With (Application) | To connect an electrofusion/electroporation system to the electrodes and chambers to be used for a particular application |
| For Use With (Equipment) | Adaptor to connect banana cables to pin tip electrodes |
| Product Line | BTX Electroporation |
| Product Type | Electroporation accessory |
| Description | Electrode Adapter Set Banana to Pin Tip |
| Display | LCD |
| Length (English) Cable | 10 ft. |
| Length (Metric) Cable | 3.1 m |
| Sterility | Non-sterile |
| Connection Type | Banana to Pin Tip |
| Show More |