Learn More
BD Vacutainer Eclipse™ Signal™ Blood Collection Needles
Product Code. 15898454
Change view
Click to view available options
Gauge:
22 G
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gauge Unit Size
15898454 22 G Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15898454 Supplier BD Vacutainer Supplier No. 368838
 Restricted Item
Estimated Shipment: 29-12-2026
Combination of proven, robust safety technology with improved in-vein confirmation.
  • Combination of proven, robust safety technology with the additional benefit of improved in-vein confirmation (speed and visibility of flash) resulting in ease of use and confidence during venous blood collection.
  • Minimizes the risk of needle stick injuries during blood collection, thereby increasing both healthcare worker and patient safety.
  • Fastest flow among BD straight needles.
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Gauge 22 G
Length (Metric) 25 mm