Learn More
Bellingham + Stanley™ Eclipse Hand Held Refractometer
GREENER_CHOICE
Product Code. 12353519
Click to view available options
Scale Range:
0 to 15 Brix
0 to 30 Brix
0 to 50 Brix
28 to 65 Brix
45 to 80 Brix
72 to 95 Brix
Increments:
0.1
0.2
0.5
Scale Type:
Brix
Low Volume Brix
Refractive Index
Unit Size:
Each
Product Code. 12353519
Brand: Bellingham + Stanley™ 4505
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Manufactured to the highest specification using only the best materials and optical components. Bellingham + Stanley™ Eclipse Hand Held Refractometer is a low-cost, portable instrument used for on-the-spot determination of Refractive Index of liquids or the concentration of solutions.
The Eclipse professional-grade range of scales are available for general use in a variety of applications, with special models available, such as Low Volume Brix models to facilitate measurement of samples of less than 1μL that are used by botanists and zoologists for measuring nectar. All instruments are supplied with a soft carrying case and instructions for use.
These reliable and robust refractometers are sealed against the ingress of water with IP65 construction, so they can be used in the harshest of environments. The large body housing and rubber handgrip make the instrument incredible easy to handle. Special features make measurements easy and reliable:
  • Simple zero adjust with lock
  • Shaded view with bright display for maximum distinction
  • High precision, clear scale
  • Easy-clean prism
  • Unique sample ‘dribble feed’ feature
  • Low weight
  • Anti-roll supports
  • Push-on prism flap
TRUSTED_SUSTAINABILITY
Specifications
Type Eclipse Professional Grade Optical Hand Held Refractometer
Scale Range 45 to 80 Brix
Increments 0.2
Scale Type Brix
For Use With (Application) ‘On the spot’ determination of Refractive Index (RI) of liquids or the concentration of solutions
Includes Soft carrying case
IP Rating IP65