Learn More
BD Eclipse™ Hypodermic Injection Needle with SmartSlip™ Technology
Description
- Unique SmartSlip technology helps to ensure a secure connection to Luer slip syringes
- Bevel oriented to safety cover accommodates low angle injections
- One technology for skin injection and blood collection
- Requires minimal change in injection technique
- The hub incorporates a plastic clip to encourage user to attach needle more securely to Luer slip syringe
- Wide textured finger pad provides safe activation area for thumb or forefinger
- Integral safety cover and extended sidewall to enhance protection
- Needle remains locked inside activated needle cover
- Activates with a touch of a finger – no hard surface required for activation
- Colour coded hub for easy needle size identification
Specifications
Specifications
| Certifications/Compliance | EU directive |
| Color | Blue |
| Needle Gauge | 23 G |
| Diameter (Metric) Outer Needle | 0.6 mm |
| For Use With (Application) | Hypodermic injections |
| Includes | Safety cover |
| Needle Point Style | Bevel |
| For Use With (Equipment) | Luer Slip Syringe |
| Connection Type | Luer |