Learn More
Labcon™ Eclipse™ 250 μL Pipette Tips with UltraFine™ Point
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bagged
Individually Wrapped
Rack, 96
Refill
Unit Size:
Pack of 1000
Pack of 200
Pack of 960
Description
- Fits popular 250 μL pipettes
- If Sterile, Validated Sterilization (ISO 11137) for use in Research and Medical Labs
- ATP, RNase/DNase and Endotoxin-free (non-pyrogenic)
- Made in USA
Specifications
Specifications
| Volume (Metric) | 250 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Length (Metric) | 52.1 mm |
| Certifications/Compliance | ISO 9001:2015, FDA CFR21 GMP, USP Class VI |
| Clean Claims | ATP, RNase/Dnase, Endotoxin-free |
| For Use With (Application) | Micropipetting |
| Format | Rack, 96 |
| Show More |