missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Echothiophate Iodide, TRC
Specifications
Specifications
| Chemical Name or Material | Echothiophate iodide |
| CAS | 513-10-0 |
| Molecular Formula | C9 H23 N O3 P S . I |
| Quantity | 100 mg |
| Synonym | Ethanaminium, 2-[(diethoxyphosphinyl)thio]-N,N,N-trimethyl-, iodide (1:1), (2-Mercaptoethyl)trimethylammonium iodide, O,O-diethyl phosphorothioate (6CI), (2-Mercaptoethyl)trimethylammonium iodide, S-ester with O,O-diethyl phosphorothioate (7CI), Ammonium, (2-mercaptoethyl)trimethyl-, iodide, S-ester with O,O-diethyl phosphorothioate (8CI), Ethanaminium, 2-[(diethoxyphosphinyl)thio]-N,N,N-trimethyl-, iodide (9CI), 217MI, Diethoxyphosphinylthiocholine iodide, Diethylphosphorylthiocholine iodide, Echothiopate iodide, Echothiophate iodide, Ecostigmine iodide, Ecostigmini jodidum, Ecothiopate iodide, N-[2-(Diethoxyphosphinylthio)ethyl]trimethylammonium iodide, Phospholine iodide, S-(2-Dimethylaminoethyl) O,O-diethylphosphorothioate methiodide, S-(Diethyl phosphono)thiocholine iodide |
| InChI Formula | InChI=1S/C9H23NO3PS.HI/c1-6-12-14(11,13-7-2)15-9-8-10(3,4)5;/h6-9H2,1-5H3;1H/q+1;/p-1 |
| SMILES | [I-].CCOP(=O)(OCC)SCC[N+](C)(C)C |
| IUPAC Name | 2-diethoxyphosphorylsulfanylethyl(trimethyl)azanium;iodide |
| Molecular Weight (g/mol) | 383.23 |
| Formula Weight | 383.0181 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?