Learn More
Terumo™ Eccentric Luer-Slip Syringe
Product Code. 17142724
Click to view available options
Volume (Metric):
10 mL
20 mL
Unit Size:
Pack of 100
Pack of 50
Product Code. 17142724
Brand: Terumo™ 8MDSS10SE
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Manufactured with polypropylene for superior thermal and chemical resistance. Terumo™ Eccentric Luer-Slip Syringes have a unique bevel design to facilitate the penetration of the skin and minimize trauma.
  • Clear polypropylene barrel and plunger; double contact ring
  • Integrated plunger stop prevents accidental pull-out
  • Sterile, Latex-free
  • Bold, precise scale markings
  • Complies to ISO 7886 (DIS 7886)
  • Individually wrapped in tamper evident peel packs
  • Supplied without needles
  • Suitable for gases or liquids
  • Eccentric luer-slip
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 7886 (DIS 7886)
Material Polypropylene
For Use With (Application) For Research Use, for Gases or Liquids
Disposable Single-use
Graduations 0.2 mL
Volume (Metric) 10 mL
Connection Type Eccentric Luer Slip
Graduated Yes