Learn More
Trajan Scientific and Medical™ EC Science Silicon Septa
Designed for long lifetime and low injection port adhesion.
Brand:  Trajan Scientific and Medical™ 041906
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15248623
  • 61.70€ / Pack of 24
Estimated Shipment: 13-06-2024
Description
  • Combines long injection lifetime, low bleed and low injection port adhesion
  • Reliable seal against carrier gas pressure in the inlet
  • No bleed or contamination into the GC system
  • Maximum operating temperature of 350°C (300°C for 17mm size)
Specifications
Specifications
EC
12.5 mm
24/Pk.