Learn More
Macherey-Nagel™ EC Analytical HPLC Column
Brand: Macherey-Nagel™ 720046.40
Additional Details : Weight : 0.01000kg
Specifications
BSA | |
4 mm | |
For HPLC, GC,TLC, SPE Application | |
5800 psi (400 bar) | |
Spherical | |
5 to 8 | |
100 m²/g | |
L75 | |
Silica | |
No | |
Analytical EC Column |
Analytical Column | |
0.7 mL/min | |
150 mm | |
50 mmol/L/L phosphate buffer + 1 % 1-propanol | |
7 μm | |
300 Å | |
40°C | |
BSA-7 | |
Mixed Mode | |
HIC, Polar Selectivity, Steric Selectivity |