missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ebio1
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Ebio1 is an activator of KCNQ2 voltage-gated potassium channels (EC50 = 247 nM). Ebio1 activates KCNQ2 by increasing both the open probability and the conductance of the channel. Ebio1 has no effect on the voltage sensitivity of KCNQ1, KCNQ3, KCNQ4 and KCNQ5.
Specifications
Specifications
| Chemical Name or Material | N-(1,2-Dihydro-5-acenaphthylenyl)-4-fluorobenzamide |
| CAS | 339287-36-4 |
| Quantity | 50 mg |
| Target | Voltage-gated Potassium Channel Activators |
| Molecular Formula | C19H14FNO |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction