Learn More
Ebro Electronic™ EBI 20 Data Logger Series
Product Code. 10272640
Click to view available options
Alarm Types:
Flashing LCD Segment
LED Indicator
Channels:
1
2
Data Memory:
20,000/Channel
40,000 Readings
IP Rating:
IP52
IP67
Unit Size:
Each
Product Code. 10272640
Brand: Ebro Electronic™ 16010044A02
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Data logger to measure temperature and / or humidity during transportation and in warehouses
EBI 20 data loggers are very robust and are in use all over the world in transportation and for warehouse monitoring.
TRUSTED_SUSTAINABILITY
Specifications
Type Datalogger
Display Type LCD
Data Memory 20,000/Channel
Calibration Calibrated
Alarm Types LED Indicator
Software Type Universal Software
Battery Type Lithium
Battery Life 24 Months
Channels 2
Certifications/Compliance Factory calibration certificate
Interface USB
IP Rating IP52
Model EBI 20-TH1
Sensor Types NTC
Accuracy (Relative Humidity) ±3%
Accuracy (Temperature) Humidity ± 3% (at 10-90% relative humidity)
Temperature ± 0.5°C (-20° to +40°C), ± 0.8°C for the remaining measuring range
Time Accuracy 1 min. to 24 hr.
Humidity Range 10 to 90%
Resolution (Relative Humidity) 0.1%
Temperature Resolution 0.5°C
Temperature Range (Metric) -30°C to +60°C
Operating Temperature (Metric) -30°C to +60°C
Dimensions (L x W x H) 69 x 48 x 22 mm
Length (Metric) 69 mm