Learn More
STEMCELL Technologies EasySep™ Multistand with 'The Big Easy' EasySep™ Magnets
Product Code. 17107851
Encompass_Preferred
Click to view available options
Includes:
4 Magnets
Unit Size:
1 set
Product Code. 17107851
Brand: STEMCELL Technologies 18100
Immunomagnetic column-free magnet
  • The EasySep™ Multistand allows the separation of up to 4 samples at one time
  • An ingenious reversible clamping bar allows the Multistand to hold up to 4 EasySep™ (Catalog #18000) or 4 'The Big Easy' EasySep™ Magnets (Catalog #18001)
  • After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes
  • The receiving tube bar is indexed to ensure proper position of the tubes
Specifications
Includes 4 Magnets
For Use With (Application) Cell Isolation
Quantity 1 Ea.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.