Learn More
STEMCELL Technologies EasySep™ Multistand with 'The Big Easy' EasySep™ Magnets
Click to view available options
Includes:
4 Magnets
Unit Size:
1 set
Description
- The EasySep™ Multistand allows the separation of up to 4 samples at one time
- An ingenious reversible clamping bar allows the Multistand to hold up to 4 EasySep™ (Catalog #18000) or 4 'The Big Easy' EasySep™ Magnets (Catalog #18001)
- After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes
- The receiving tube bar is indexed to ensure proper position of the tubes
Specifications
Specifications
| Includes | 4 Magnets |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 Ea. |