missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Mouse SCA1 Positive Selection Kit
Click to view available options
Quantity:
2 x 109 cells
Unit Size:
1 set
Description
- The EasySep™ Mouse SCA1 Positive Selection Kit is designed to isolate SCA1+ cells from single-cell suspensions of bone marrow by positive selection
- The cocktail contains a combination of PE-labeled antibodies that is directed against SCA1, which are then recognized by antibody complexes that are directed against PE and dextran
- Labeled cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- This kit replaces the EasySep™ Mouse SCA1 Biotin Positive Selection Kit (Catalog #18856) for even faster cell isolations
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 2 x 109 cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction