missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human Progenitor Cell Enrichment Kit with Platelet Depletion
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
Description
- The EasySep™ Human Progenitor Cell Enrichment Kit with Platelet Depletion is designed to isolate hematopoietic progenitor cells from fresh or previously frozen cord blood and other cell preparations that contain large numbers of platelets by negative selection
- Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD3, CD11b, CD11c, CD14, CD16, CD19, CD24, CD56, CD61, CD66b, glycophorin A and dextran-coated magnetic particles
- The labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction