missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Human NK Cell Enrichment Kit

Product Code. 15877821
Encompass
Change view
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
Product Code. Quantity unitSize
15877821 1 x 109 cells 1 set
1 options
This item is not returnable. View return policy

Product Code. 15877821

Brand: STEMCELL Technologies 19055

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic isolation of untouched human NK cells

  • The EasySep™ Human NK Cell Enrichment Kit is designed to isolate NK cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-NK cells and dextran-coated magnetic particles
  • The labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube

Specifications

Cell Type NK Cells
For Use With (Application) Cell Isolation
Quantity 1 x 109 cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.