missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human Monocyte Enrichment Kit without CD16 Depletion
Description
- The EasySep™ Human Monocyte Enrichment Kit without CD16 Depletion is designed to isolate monocytes and CD14+CD16+ monocytes from fresh or previously frozen peripheral blood mononuclear cells by negative selection
- The CD14+CD16+ subset of monocytes (∼10% in blood of healthy individuals) has characteristics of tissue macrophages and expands greatly in acute and chronic inflammatory disease
- Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-monocyte cells and magnetic particles
- The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
- The labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
Specifications
Specifications
| Cell Type | Monocytes |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 Cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?