missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Human Memory B Cell Isolation Kit

Product Code. 15845427
Encompass
Click to view available options
Quantity:
2 x 109 cells
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15845427

Brand: STEMCELL Technologies 17864

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic positive selection of human memory B cells

  • The EasySep™ Human Memory B Cell Isolation Kit is a two-step method designed to isolate CD27+ B cells from fresh or previously frozen peripheral blood mononuclear cells
  • First, CD27+ cells are isolated by column-free immunomagnetic positive selection using antibody complexes and EasySep™ Releasable RapidSpheres™
  • Then, bound magnetic particles are removed from the EasySep™-isolated CD27+ cells, and unwanted non-B cells are targeted for depletion using antibody complexes and EasySep™ Dextran RapidSpheres™
  • The final isolated fraction contains highly purified CD27+ memory B cells that are immediately ready for downstream applications
  • An optional protocol allows for the isolation of naïve B cells in parallel for use in functional studies

Specifications

Cell Type B Cells
For Use With (Application) Cell Isolation
Quantity 2 x 109 cells
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.