missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Human Extracellular Vesicle (CD63) Positive Selection Kit

Product Code. 17167811
Encompass
Change view
Click to view available options
Quantity:
20 mL of biofluid
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17167811 20 mL of biofluid 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17167811 Supplier STEMCELL Technologies Supplier No. 17895

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fast & easy isolation of human extracellular vesicles using immunomagnetic positive selection

  • The EasySep™ Human Extracellular Vesicle (CD63) Positive Selection Kit isolates highly purified human extracellular vesicles from plasma/serum and cell culture conditioned medium by immunomagnetic positive selection
  • Extracellular vesicles are labeled with Tetrameric Antibody Complexes recognizing CD63 and magnetic particles
  • Labeled extracellular vesicles are separated without columns using an EasySep™ magnet
  • Unwanted biofluid components are simply poured off, while desired extracellular vesicles remain in the tube
  • Isolated extracellular vesicles are available for downstream applications such as DNA/RNA extraction, Western blot, or mass spectometry​

Specifications

Cell Type Other
For Use With (Application) Cell Isolation
Quantity 20 mL of biofluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.