missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human Eosinophil Isolation Kit
Description
- The EasySep™ Human Eosinophil Isolation Kit is designed to isolate eosinophils from a polymorphonuclear cell-rich fraction of peripheral blood by negative selection
- Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-eosinophils and magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
- The kit is compatible with cells prepared using either Lymphoprep™ (Catalog #07801) centrifugation (preferred for higher cell purities) or HetaSep™ (Catalog #07906) sedimentation (preferred for faster, lysis-free sample processing)
- This product replaces the EasySep™ Human Eosinophil Enrichment Kit (Catalog #19256) for even faster cell isolations
Specifications
Specifications
| Cell Type | Granulocytes and Subsets |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?