missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human CD4 Positive Selection Kit II
Description
- The EasySep™ Human CD4 Positive Selection Kit II is designed to isolate CD4+ cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic positive selection
- Desired cells are targeted with antibody complexes recognizing CD4 and magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- This product replaces the EasySep™ Human CD4 Positive Selection Kit (Catalog #18052) for even faster cell isolations
Specifications
Specifications
| Cell Type | T Cells, T Cells, CD4+ |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?