missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Human CD2 Positive Selection Kit II
Click to view available options
Quantity:
1 x 109 Cells
Unit Size:
1 set
Description
- The EasySep™ Human CD2 Positive Selection Kit II is designed to isolate CD2+ cells from fresh or previously frozen human peripheral blood mononuclear cells by positive selection
- Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD2 and dextran-coated magnetic particles
- The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- The CD2 antigen is expressed on T cells and NK cells
- This product replaces the EasySep™ Human CD2 Positive Selection Kit (Catalog #18657) for even faster cell isolations
Specifications
Specifications
| Cell Type | T Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 Cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction